Merck
  • Selective PRMT5 Inhibitors Suppress Human CD8+ T Cells by Upregulation of p53 and Impairment of the AKT Pathway Similar to the Tumor Metabolite MTA.
Selective PRMT5 Inhibitors Suppress Human CD8+ T Cells by Upregulation of p53 and Impairment of the AKT Pathway Similar to the Tumor Metabolite MTA.
Molecular cancer therapeutics (2019-11-13)
Carolin Dorothea Strobl, Stefanie Schaffer, Tabea Haug, Simon Völkl, Katrin Peter, Katrin Singer, Martin Böttcher, Dimitrios Mougiakakos, Andreas Mackensen, Michael Aigner
ABSTRACT
Genetic alterations in tumor cells provide promising targets for antitumor therapy. Recently, loss of methylthioadenosine phosphorylase (MTAP), a deletion frequently occurring in cancer, has been shown to create vulnerability to the inhibition of the protein arginine methyltransferase 5 (PRMT5). MTAP deficiency leads to accumulation of methylthioadenosine (MTA), which reduces PRMT5 activity, and thus, sensitizes the tumor cells to selective PRMT5 inhibitors (PRMT5i). PRMT5i are investigated as a new strategy to selectively kill MTAP-deficient tumor cells by blocking residual PRMT5 activity, but also to treat PRMT5-overexpressing tumors. Although many studies investigated the role of PRMT5 in cancer, only little data exist about the effect of PRMT5 inhibition on immune cells. As we could show that the tumor metabolite MTA suppresses T cells, we asked whether selective PRMT5 inhibition is detrimental for T-cell immune responses. Therefore, we examined the effect of the synthetic PRMT5 inhibitor EPZ015666 on human CD8+ T cells in direct comparison with the naturally occurring PRMT5-inhibiting molecule MTA. Both compounds reduced T-cell proliferation, viability, and functionality. In addition, T-cell metabolism was impaired upon PRMT5 inhibition. These effects coincided with the induction of p53 expression and reduced AKT/mTOR signaling. Our data clearly demonstrate that PRMT5 activity is involved in various cellular processes of human CD8+ T cells associated with essential T-cell functions. Therefore, not only tumor cells, but also antitumor immune responses, are compromised by PRMT5 inhibitors. This emphasizes the importance of considering side effects on the immune system when developing new strategies to specifically target not only MTAP-deficient tumors.
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5′-Deoxy-5′-(methylthio)adenosine
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
EPZ015666, ≥98% (HPLC)
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Ionomycin calcium salt, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO