Merck
  • Understanding the interaction between α-1-acid glycoprotein (AGP) and potential Cu/Zn metallo-drugs of benzimidazole derived organic motifs: A multi-spectroscopic and molecular docking study.
Understanding the interaction between α-1-acid glycoprotein (AGP) and potential Cu/Zn metallo-drugs of benzimidazole derived organic motifs: A multi-spectroscopic and molecular docking study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2019-08-27)
Mohamed F AlAjmi, Md Tabish Rehman, Rais Ahmad Khan, Meraj A Khan, Ghazala Muteeb, Mohd Shahnawaz Khan, Omar Mohammed Noman, Ali Alsalme, Afzal Hussain
ABSTRACT
Drug-binding and interactions with plasma proteins strongly affect their efficiency of delivery, hence considered as a key factor in determining the overall pharmacological action. Alpha-1-acid glycoprotein (AGP), a second most abundant plasma protein in blood circulation, has unique drug binding ability and involved in the transportation of various compounds. Here, we have investigated the mechanism of interaction between AGP and potential Cu/Zn metallo-drugs of benzimidazole derived organic motifs (CuL2 and ZnL2, where L is Schiff base ligand) by applying integrated spectroscopic, biophysical techniques and computational molecular docking analyses. We found that both the metallo-drugs (CuL2 and ZnL2) were bound at the central cavity of AGP interacting with the residues of lobe I, lobe II as well as lobe III. The binding of metallo-drugs to AGP occurs in 1:1 M ratios. Hydrogen bonding, electrostatic and hydrophobic interactions played a significant role in stabilizing the AGP-metallo-drug complexes. Binding affinities of both the metallo-drugs towards AGP at 298 K were of the order of 104-105 M-1, corresponding to Gibbs free energy of stabilization of approximately -5.50 to -6.62 kcal mol-1. Furthermore, the spectroscopic investigation by circular dichroism and synchronous fluorescence analyses suggest conformational changes in AGP upon the binding of metallic compounds.
Sigma-Aldrich
α1-Acid Glycoprotein from bovine plasma, ≥99%