Merck
  • Comparing the release of erythromycin and vancomycin from calcium polyphosphate hydrogel using different drug loading methods.
Comparing the release of erythromycin and vancomycin from calcium polyphosphate hydrogel using different drug loading methods.
Journal of biomedical materials research. Part B, Applied biomaterials (2019-05-10)
Yasaman Chehreghanianzabi, Rajib Barua, Tong Shi, Sally Yurgelevic, Gregory Auner, David C Markel, Weiping Ren
ABSTRACT
Calcium polyphosphate (CPP) hydrogel is used to load erythromycin (EM) and vancomycin (VCM) by means of two loading methods: they are either added directly to the formed CPP hydrogel (Gel Mixture method) or mixed with CPP powders, followed by the formation of CPP-antibiotic hydrogel (Powder Mixture method). The release of loaded antibiotics from CPP hydrogel is measured up to 48 hr. Compared to Powder Mixture method, Gel Mixture method significantly reduced the burst release of embedded antibiotics. A significant change in CPP hydrogel Raman characteristic peaks is observed only in Gel Mixture method, indicating a close interaction between embedded antibiotics with CPP hydrogel matrix. In contrast, a similarity between characteristic peaks of CPP hydrogel and Powder Mixture method shows that antibiotic incorporation does not interfere with CPP gel formation, resulting in no ionic interaction between antibiotic and polyphosphate chains. Rheometer analysis further confirms that the hydrophobic nature of EM impacts the viscoelastic properties of CPP hydrogel, whereas the hydrophilic VCM exhibits a higher loading efficiency. The potential application of CPP hydrogel as a ceramic matrix for sustained drug release warrants further investigation.
Sigma-Aldrich
Calcium phosphate monobasic, ≥95%