Merck
  • Mucin 13: structure, function, and potential roles in cancer pathogenesis.
Mucin 13: structure, function, and potential roles in cancer pathogenesis.
Molecular cancer research : MCR (2011-04-01)
Diane M Maher, Brij K Gupta, Satoshi Nagata, Meena Jaggi, Subhash C Chauhan
ABSTRACT
Mucin 13 (MUC13) is a high-molecular-weight transmembrane glycoprotein that is frequently and aberrantly expressed in a variety of epithelial carcinomas, including gastric, colorectal, and ovarian cancers. On the basis of the high expression of MUC13 in cancer cells as well as recent laboratory findings suggesting a malignant phenotype of MUC13-transfected cell lines, the oncogenic potential of MUC13 has emerged. The various functional domains of MUC13 may confer oncogenic potential to MUC13. For example, the bulky extracellular domain with extensive modification with glycan chains may prevent cell-cell and cell-extracellular matrix binding whereas the cytoplasmic tail containing serine and tyrosine residues for potential phosphorylation may participate in cell signaling. MUC13 exhibits the characteristics suitable as an early marker for cancer screening and presents a promising target for antibody-guided targeted therapy.