Merck
  • The calcium-sensitive large-conductance potassium channel (BK/MAXI K) is present in the inner mitochondrial membrane of rat brain.
The calcium-sensitive large-conductance potassium channel (BK/MAXI K) is present in the inner mitochondrial membrane of rat brain.
Neuroscience (2006-03-29)
R M Douglas, J C K Lai, S Bian, L Cummins, E Moczydlowski, G G Haddad
ABSTRACT
Large-conductance voltage- and calcium-sensitive channels are known to be expressed in the plasmalemma of central neurons; however, recent data suggest that large-conductance voltage- and calcium-sensitive channels may also be present in mitochondrial membranes. To determine the subcellular localization and distribution of large-conductance voltage- and calcium-sensitive channels, rat brain fractions obtained by Ficoll-sucrose density gradient centrifugation were examined by Western blotting, immunocytochemistry and immuno-gold electron microscopy. Immunoblotting studies demonstrated the presence of a consistent signal for the alpha subunit of the large-conductance voltage- and calcium-sensitive channel in the mitochondrial fraction. Double-labeling immunofluorescence also demonstrated that large-conductance voltage- and calcium-sensitive channels are present in mitochondria and co-localize with mitochondrial-specific proteins such as the translocase of the inner membrane 23, adenine nucleotide translocator, cytochrome c oxidase or complex IV-subunit 1 and the inner mitochondrial membrane protein but do not co-localize with calnexin, an endoplasmic reticulum marker. Western blotting of discrete subcellular fractions demonstrated that cytochrome c oxidase or complex IV-subunit 1 was only expressed in the mitochondrial fraction whereas actin, acetylcholinesterase, cadherins, calnexin, 58 kDa Golgi protein, lactate dehydrogenase and microtubule-associated protein 1 were not, demonstrating the purity of the mitochondrial fraction. Electron microscopic examination of the mitochondrial pellet demonstrated gold particle labeling within mitochondria, indicative of the presence of large-conductance voltage- and calcium-sensitive channels in the inner mitochondrial membrane. These studies provide concrete morphological evidence for the existence of large-conductance voltage- and calcium-sensitive channels in mitochondria: our findings corroborate the recent electrophysiological evidence of mitochondrial large-conductance voltage- and calcium-sensitive channels in glioma and cardiac cells.