Merck
  • Succinate induces synovial angiogenesis in rheumatoid arthritis through metabolic remodeling and HIF-1α/VEGF axis.
Succinate induces synovial angiogenesis in rheumatoid arthritis through metabolic remodeling and HIF-1α/VEGF axis.
Free radical biology & medicine (2018-07-22)
Yi Li, Yang Liu, Chen Wang, Wen-Rui Xia, Jia-Yi Zheng, Jie Yang, Baolin Liu, Jian-Qun Liu, Li-Fang Liu
ABSTRACT
In response to hypoxic succinate accumulates in arthritis synovium, however, the implication is little known. This study aims to investigate whether succinate could act as a metabolic signal linking metabolic alternation with angiogenesis in arthritis synovium. The interaction between elevated succinate and VEGF production was examined in endothelial cells. Succinate production, HIF-1α induction and angiogenesis in the hypoxic synovium of collagen-induced arthritis rats were also investigated. Intracellular succinate promoted VEGF production and induced angiogenic response dependent on HIF-1α induction in endothelial cells. Luciferase reporter assay showed that succinate increased VEGF expression through gene promoter activation dependent on HIF-1α induction. Intracellular succinate released into intercellular space, where extracellular succinate activated succinate receptor G-protein-coupled receptor 91 (GPR91) and induced VEGF production, further exacerbating angiogenesis. In addition, TGF-β1 treatment increased succinate production due to the reversal of succinate dehydrogenase (SDH) activation, and consistently, SDH inhibitor dimethyl malonate reduced angiogenesis in the arthritis synovium. More than an intermediate, succinate functioned as a signaling molecule to link metabolic reprograming with angiogenesis. Intracellular succinate induced angiogenesis through HIF-1α induction, while extracellular succinate acted on GPR91 activation, working together to disturb energy metabolism and exacerbate inflammation and angiogenesis in arthritis synovium. Our work suggested that suppression of SDH could prevent succinate accumulation and inhibit angiogenesis via blocking HIF-1α/VEGF axis. This finding not only provides a novel insight into angiogenesis, but also reveals a potential therapeutical strategy to attenuate revascularization in arthritis.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
MISSION® esiRNA, targeting human SDHB