Skip to Content
Merck
Uncarine E phyproof® Reference Substance

Uncarine E

Uncarine E phyproof® Reference Substance
Synonym(s):
7-Isopteropodine
Empirical Formula (Hill Notation):
C21H24N2O4
CAS Number:
Molecular Weight:
368.43

Products

Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing