Monolayer hexagonal Boron Nitride (hBN) on copper foil
Linear Formula:
h-BN
CAS Number:
Molecular Weight:
24.82
Products
Brand
Formula Weight
Diameter (mm)
Quality Segment
Particle Size (µm)
Showing 1-5 of 5
Compare
Product Number
Description
Pricing