Skip to Content
Merck
vibrant-m

Chorionic Gonadotropin, Human Urine, Standard Grade

vibrant-m
Synonym(s):
Chorionic Gonadotropin, Human Urine, Standard Grade, Chorionic gonadotropin human, Choriogonin, HCG
CAS Number:

Products

Product Category
Brand
Color
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility