Skip to Content
Merck
L-Alanine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

L-Alanine

L-Alanine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(S)-2-Aminopropionic acid, L-α-Aminopropionic acid, L-Alanine
Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique