Skip to Content
Merck
β-Estradiol analytical standard

17β-Estradiol solution

β-Estradiol analytical standard
Synonym(s):
β-Estradiol, 1,3,5-Estratriene-3,17β-diol, 17β-Estradiol, 3,17β-Dihydroxy-1,3,5(10)-estratriene, Dihydrofolliculin
Empirical Formula (Hill Notation):
C18H24O2
CAS Number:
Molecular Weight:
272.38
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique