Catechol
Synonym(s):
Pyrocatechol, 1,2-Benzenediol, 1,2-Dihydroxybenzene, 2-Hydroxyphenol, Catechol
Linear Formula:
C6H4-1,2-(OH)2
CAS Number:
Molecular Weight:
110.11
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Color
Package Size
API Family
Reference Material Type
Special Grade
Technique