Ethylenediaminetetraacetic acid solution
Synonym(s):
EDTA solution, EDTA, 500 mM Solution, pH 8.0, ULTROL® Grade, Ethylenedinitrilotetraacetic acid
Empirical Formula (Hill Notation):
C10H16N2O8
CAS Number:
Molecular Weight:
292.24
Products
Brand
Color
Formula Weight
Physical Form
Product Line
Quality Segment
Reaction Type
Reagent Type
Sterilization Method
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing