L-α-Phosphatidylcholine
Synonym(s):
1,2-Diacyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Purity
Tissue
Quality Segment
Functional Group
Optical Isomer
Technique
Showing 1-2 of 2