Behenic Acid
Synonym(s):
Paricalcitol, (1α, 3β, 7E,22E)-19-Nor-9,10-secoergosta-5,7,22-triene-1,3,25-triol
Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
Products
Brand
Formula Weight
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing