Ascentis® RP-Amide Supelguard™ Guard Cartridge
Products
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Column Material
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing