Skip to Content
Merck
Ammonium chloride tested according to Ph. Eur.

Ammonium chloride

Ammonium chloride tested according to Ph. Eur.
Synonym(s):
Salmiac, Ammonium chloride
Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique