4′-[(1,7′-Dimethyl-2′-propyl[2,5′-bi-1H-benzimidazol]-1′-yl)methyl][1,1′-biphenyl]-2-carboxylic acid
Synonym(s):
4′-[(1,7′-Dimethyl-2′-propyl-1H,1′H-2,5′-bibenzo[d]imidazol-1′-yl)methyl]biphenyl-2-carboxylic acid
Empirical Formula (Hill Notation):
C33H30N4O2
CAS Number:
Molecular Weight:
514.62
Products
Feature
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1