MES monohydrate
Synonym(s):
2-(N-Morpholino)ethanesulfonic acid, 4-Morpholineethanesulfonic acid monohydrate, 2-Morpholinoethanesulfonic acid monohydrate, 2-Morpholinoethanesulfonic acid monohydrate (MES), OmniPur® MES, Monohydrate
Linear Formula:
(C4H8ON)CH2CH2SO3H · H2O
CAS Number:
Molecular Weight:
213.25
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing