Potassium phosphate dibasic
Synonym(s):
Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate, Potassium phosphate dibasic
Linear Formula:
K2HPO4
CAS Number:
Molecular Weight:
174.18
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-15 of 15
Compare
Product Number
Description
Pricing