L-Phenylalanine
Synonym(s):
(S)-2-Amino-3-phenylpropionic acid, L-Phenylalanine, (S)-(-)-Phenylalanine
Linear Formula:
C6H5CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
165.19
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Optical Isomer
Amino Acid
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing