Alginic acid sodium salt
Synonym(s):
Algin, Sodium alginate, Alginic acid sodium salt from brown algae
CAS Number:
Products
Product Category
Brand
Biological Source
Color
pH Value
Physical Form
Product Line
Agency
Quality Segment
Application
Organoleptic Class
Polymer Composition
Polymer Type
Special Grade
Showing 1-5 of 5