Zirconium(IV) oxide
Synonym(s):
Zirconia, Zirconium(IV) oxide
Linear Formula:
ZrO2
CAS Number:
Molecular Weight:
123.22
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Particle Size (µm)
Reagent Type
Core
Showing 1-7 of 7
Compare
Product Number
Description
Pricing