Skip to Content
Merck
All Photos(1)

Key Documents

AB9552

Sigma-Aldrich

Anti-Chemokine-Like Receptor 1 Antibody

Chemicon®, from rabbit

Synonym(s):

CMKLR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

availability

not available in Japan

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CMKLR1(1240)

Specificity

Recognizes Chemokine-Like Receptor 1 [CMKLR1].

Immunogen

Synthetic peptide from the N-terminal extracellular domain of human Chemokine-Like Receptor 1. The immunogen has 58% conservation with mouse and 63% with rat.

Application

Anti-Chemokine-Like Receptor 1 Antibody is an antibody against Chemokine-Like Receptor 1 for use in IC, IH(P).
Immunohistochemistry on formalin fixed paraffin embedded tissue sections.

Immunocytochemistry on acetone fixed cells.

Optimal working dilutions must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Physical form

Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide.

Storage and Stability

Maintain at -70°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service