Merck
  • Vancomycin-Resistant Entrococci colonization in chronic hemodialysis patients and its risk factors in southern Iran (2005-2006).
Vancomycin-Resistant Entrococci colonization in chronic hemodialysis patients and its risk factors in southern Iran (2005-2006).
Iranian Red Crescent medical journal (2013-01-04)
S Shaghaghian, B Pourabbas, A Alborzi, M Askarian, J Mardaneh
ABSTRACT
Vancomycin-resistant enterococci (VRE) recently have emerged as a nosocomial pathogen among dialysis patients. This study aims to appraise the prevalence, incidence density and risk factors for VRE colonization among these patients. In this prospective study, 782 stool or rectal swab specimens were collected from 250 chronic hemodialysis patients with an interval of at least one month. To identify the risk factors of VRE colonization, demographic and health data of VRE+ and VRE- patients were compared. VRE colonization was detected in 55 (22%) patients during study. Incidence density of one case per 79.6 patient-month of follow up was estimated.The only significant difference between the data collected from VRE+ and VRE- patients was observed in antibiotic consumption (P<0.001). VRE colonization is relatively high and rapidly spreading among chronic dialysis patients. It is strongly associated with recently antibiotic consumption.
Sigma-Aldrich
D-(+)-Sorbose, ≥99%