Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tenax
®
GR
Synonym(s):
Tenax
®
GR
Products
Brand
Supelco (5)
Sustainability
Feature
ID (mm)
OD (mm)
Compatibility
Quality Segment
Application
Package Size
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
29549-U
Tenax
®
GR,
O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, preconditioned, pkg of 10 ea
Expand
View Pricing
28272-U
Tenax
®
GR,
stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Expand
View Pricing
20011-U
Tenax
®
GR,
stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Expand
View Pricing
29742-U
Tenax
®
GR,
60/80 mesh, O.D. × L 1/4 in. × 3 1/2, stainless steel TD tube, sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Expand
View Pricing
28282-U
Tenax
®
GR,
glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Expand
View Pricing