Skip to Content
Merck
DL-α-Tocopherol acetate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Tocopheryl Acetate, a

DL-α-Tocopherol acetate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
DL-α-Tocopherol acetate, Vitamin E acetate, all-rac-α-Tocopheryl acetate, O-Acetyl-α-tocopherol, DL-α-Tocopheryl acetate
Empirical Formula (Hill Notation):
C31H52O3
CAS Number:
Molecular Weight:
472.74
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Technique