Skip to Content
Merck
vibrant-m

Dyglycine

vibrant-m
Synonym(s):
N-Glycylglycine
Empirical Formula (Hill Notation):
C4H8N2O3
CAS Number:
Molecular Weight:
132.12

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade