L-Valine
Synonym(s):
L-Valine, (S)-α-Aminoisovaleric acid, L-2-Amino-3-methylbutanoic acid
Linear Formula:
(CH3)2CHCH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing