Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Boric acid
Linear Formula:
H
3
BO
3
CAS Number:
10043-35-3
Molecular Weight:
61.83
EC Number:
233-139-2
Products
Product Category
buffers (6)
inorganic acids (3)
salts (1)
Brand
Sigma-Aldrich (10)
Supelco (2)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (1)
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
B0394
Boric acid,
ACS reagent, ≥99.5%
Expand
View Pricing
B6768
Boric acid,
BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Expand
View Pricing
B0252
Boric acid,
ReagentPlus
®
, ≥99.5%
Expand
View Pricing
1.00165
Boric acid,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
31146
Boric acid,
puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
Expand
View Pricing
B7901
Boric acid,
suitable for electrophoresis, ≥99.5%
Expand
View Pricing
15663
Boric acid,
BioUltra, for molecular biology, ≥99.5% (T)
Expand
View Pricing
202878
Boric acid,
99.999% trace metals basis
Expand
View Pricing
339067
Boric acid,
99.97% trace metals basis
Expand
View Pricing
11607
Boric acid,
puriss., meets analytical specification of Ph. Eur., BP, NF, 99.5-100.5%, powder
Expand
View Pricing
B7660
Boric acid,
BioXtra, ≥99.5%
Expand
View Pricing
1.00765
Boric acid,
99.9999 Suprapur
®
Expand
View Pricing
B2645
Boric acid,
tablet, 1 g boric acid per tablet
Expand
View Pricing