Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Trifluoroacetic acid
Synonym(s):
TFA
Linear Formula:
CF
3
COOH
CAS Number:
76-05-1
Molecular Weight:
114.02
EC Number:
200-929-3
Products
Product Category
solvents (4)
organic acids (2)
buffers (1)
Brand
Sigma-Aldrich (5)
Supelco (4)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Anhydrous (1)
Puriss (1)
analytical standard (1)
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
T6508
Trifluoroacetic acid,
ReagentPlus
®
, 99%
Expand
View Pricing
302031
Trifluoroacetic acid,
suitable for HPLC, ≥99.0%
Expand
View Pricing
91707
Trifluoroacetic acid,
puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Expand
View Pricing
8.08260
Trifluoroacetic acid,
for synthesis
Expand
View Pricing
1.08262
Trifluoroacetic acid,
for spectroscopy Uvasol
®
Expand
View Pricing
299537
Trifluoroacetic acid,
≥99%, for protein sequencing
Expand
View Pricing
80457
Trifluoroacetic acid,
eluent additive for LC-MS, LiChropur
™
, ≥99.0% (GC)
Expand
View Pricing
1.08178
Trifluoroacetic acid,
for protein sequence analysis
Expand
View Pricing
74564
Trifluoroacetic acid,
analytical standard
Expand
View Pricing
900518
Trifluoroacetic acid,
anhydrous, ZerO
2
®
, ≥99.0%
Expand
View Pricing