Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Chlorobenzene
Synonym(s):
Chlorobenzene
Empirical Formula (Hill Notation):
C
6
H
5
Cl
CAS Number:
108-90-7
Molecular Weight:
112.56
EC Number:
203-628-5
Products
Product Category
solvents (6)
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Anhydrous (2)
ACS reagent (1)
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
284513
Chlorobenzene,
anhydrous, 99.8%
Expand
View Pricing
101389
Chlorobenzene,
ReagentPlus
®
, 99%
Expand
View Pricing
270644
Chlorobenzene,
suitable for HPLC, 99.9%
Expand
View Pricing
319996
Chlorobenzene,
ACS reagent, ≥99.5%
Expand
View Pricing
8.01791
Chlorobenzene,
for synthesis
Expand
View Pricing
8.14506
Chlorobenzene,
special quality for synthesis
Expand
View Pricing
900521
Chlorobenzene,
anhydrous, ZerO
2
®
, 99.8%
Expand
View Pricing