Skip to Content
Merck
All Photos(5)

Key Documents

WH0004723M1

Sigma-Aldrich

Monoclonal Anti-NDUFV1 antibody produced in mouse

clone 4A7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa, Anti-UQOR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDUFV1(4723)

Related Categories

General description

The NDUFV1 gene encodes the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.[supplied by OMIM

Immunogen

NDUFV1 (NP_002466, 365 a.a. ~ 464 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
KAIARLIEFYKHESCGQCTPCREGVDWMNKVMARFVRGDARPAEIDSLWEISKQIEGHTICALGDGAAWPVQGLIRHFRPELEERMQRFAQQHQARQAAS

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service