Skip to Content
Merck
All Photos(1)

Key Documents

49673

Supelco

Dibenzyl ether

analytical standard

Synonym(s):

Benzyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€348.00

€348.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€348.00

About This Item

Linear Formula:
(C6H5CH2)2O
CAS Number:
Molecular Weight:
198.26
Beilstein:
1911156
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€348.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.561-1.563

bp

158-160 °C/10 mmHg (lit.)

mp

1.5-3.5 °C (lit.)

density

1.043 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

278.6 °F - Pensky-Martens closed cup

Flash Point(C)

137 °C - Pensky-Martens closed cup


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Jin Han et al.
    ACS nano, 9(3), 2805-2819 (2015-02-18)
    Electrophysiological phenotype development and paracrine action of mesenchymal stem cells (MSCs) are the critical factors that determine the therapeutic efficacy of MSCs for myocardial infarction (MI). In such respect, coculture of MSCs with cardiac cells has windowed a platform for
    Lijiao Ao et al.
    Nanoscale, 6(18), 10710-10716 (2014-08-07)
    This paper devotes a novel micellar structure for cancer theranostics by incorporating magnetic and therapeutic functionalities into a natural sourced targeting polymer vehicle. Heparin-folic acid micelles taking advantage of both excellent loading capability and cancer targeting ability have been employed
    Benjamin Matt et al.
    Angewandte Chemie (International ed. in English), 53(46), 12446-12450 (2014-09-10)
    Organizing magnetic nanoparticles into long-range and dynamic assemblies would not only provide new insights into physical phenomena but also open opportunities for a wide spectrum of applications. In particular, a major challenge consists of the development of nanoparticle-based materials for
    Dan Heo et al.
    Nanotechnology, 25(27), 275102-275102 (2014-06-25)
    In this study, we developed the maleimidyl magnetic nanoplatform, which enables functional targeting of a biomarker-specific moiety for molecular imaging via MRI. The maleimide group of the maleimidyl magnetic nanoplatform is conjugated with a thiol group without additional crosslinkers and
    Sasmita Mohapatra et al.
    Dalton transactions (Cambridge, England : 2003), 43(42), 15841-15850 (2014-09-17)
    The development of theranostic mesoporous hollow silica nanospheres having therapeutic and diagnostic functions has been achieved. The exterior surface of the hollow mesoporous silica nanosphere was selectively acid functionalized and utilised to conjugate the anticancer drug cisplatin, the marker molecule

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service