Skip to Content
Merck
All Photos(1)

Key Documents

MABF142A

Sigma-Aldrich

Anti-Ly-6G (mouse), APC-Cy7, clone 1A8 Antibody

clone 1A8, from rat, APC-Cy7

Synonym(s):

Lymphocyte antigen 6G (Ly-6G), Ly-6G.1, Ly-6G

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

APC-Cy7

antibody form

purified antibody

antibody product type

primary antibodies

clone

1A8, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Ly6G(546644)

General description

The 1A8 antibody binds to mouse Ly-6G, commonly known as Gr-1, a member of the Ly-6 superfamily of GPI-anchored cell surface proteins with roles in cell signaling and cell adhesion. Gr-1 is differentially expressed during development and maturation of cells in the myeloid lineage and is expression at varying stages and levels on monocytes, macrophages, granulocytes, and peripheral neutrophils. In the mouse, the 1A8 antibody is typically used in combination with the macrophage labeling antibody M1/70 (Anti-CD11b) for phenotypic analysis of monocytes, macrophages and granulocytes.

Application

This Anti-Ly-6G (mouse), APC-Cy7, clone 1A8 Antibody is validated for use in Flow Cytometry for the detection of Ly-6G.

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.5 μg from a representative lot detected Ly-6G in one million C57Bl/6 bone marrow cells.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service