Skip to Content
Merck
All Photos(1)

Key Documents

411795

Sigma-Aldrich

Vinyl decanoate

contains monomethyl ether hydroquinone as inhibitor, >99%

Synonym(s):

Vinyl n -decanoate, Vinyl caprate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CO2CH=CH2
CAS Number:
Molecular Weight:
198.30
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

>99%

form

liquid

contains

monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.435 (lit.)

bp

119-120 °C/10 mmHg (lit.)

density

0.886 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC(=O)OC=C

InChI

1S/C12H22O2/c1-3-5-6-7-8-9-10-11-12(13)14-4-2/h4H,2-3,5-11H2,1H3

InChI key

CMDXMIHZUJPRHG-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanhong Bi et al.
Frontiers in bioengineering and biotechnology, 8, 485-485 (2020-06-18)
The immobilization of Thermomyces lanuginosus lipase on polydopamine-functionalized Fe3O4 magnetic nanoparticles (Fe3O4@PDA-TLL) as a nanobiocatalyst was successfully performed for the first time, and the Fe3O4@PDA-TLL was used for regioselective acylation of natural hyperoside with vinyl decanoate. The effects of several
Rebecca Hollenbach et al.
Frontiers in bioengineering and biotechnology, 8, 382-382 (2020-05-21)
Glycolipids are considered an alternative to petrochemically based surfactants because they are non-toxic, biodegradable, and less harmful to the environment while having comparable surface-active properties. They can be produced chemically or enzymatically in organic solvents or in deep eutectic solvents
Pranee Inprakhon et al.
Journal of biotechnology, 259, 182-190 (2017-07-29)
Sucrose monocaprate was synthesized by carrying out a lipase-catalyzed transesterification in a non-aqueous biphasic medium. Vinyl caprate was mechanically dispersed into a solution of sucrose in DMSO. The use of DMSO allowed increasing sucrose concentration up to 0.7M (in DMSO).
Xuan Li et al.
Journal of colloid and interface science, 556, 616-627 (2019-09-09)
The development of functional and nutritional surfactants for the food industry remains a subject of great interest. Herein, therefore, we report on the design and synthesis of novel trisaccharide (raffinose) monoester-based surfactants in the expectation that they would display functional
Xi Zhang et al.
Journal of the science of food and agriculture, 95(8), 1631-1637 (2014-08-12)
Sugar medium-chain fatty acid esters are a new type of biodegradable, non-toxic, non-irritant and non-ionic surfactant with proven antimicrobial activity. Various sugar medium-chain fatty monoesters were prepared enzymatically using Lipozyme TLIM in organic solvent. Properties such as surface tension, antimicrobial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service