Merck
  • Lysophosphatidic acid via LPA-receptor 5/protein kinase D-dependent pathways induces a motile and pro-inflammatory microglial phenotype.
Lysophosphatidic acid via LPA-receptor 5/protein kinase D-dependent pathways induces a motile and pro-inflammatory microglial phenotype.
Journal of neuroinflammation (2017-12-21)
I Plastira, E Bernhart, M Goeritzer, T DeVaney, H Reicher, A Hammer, B Lohberger, A Wintersperger, B Zucol, W F Graier, D Kratky, E Malle, W Sattler
ABSTRACT
Extracellular lysophosphatidic acid (LPA) species transmit signals via six different G protein-coupled receptors (LPAR1-6) and are indispensible for brain development and function of the nervous system. However, under neuroinflammatory conditions or brain damage, LPA levels increase, thereby inducing signaling cascades that counteract brain function. We describe a critical role for 1-oleyl-2-hydroxy-sn-glycero-3-phosphate (termed "LPA" throughout our study) in mediating a motile and pro-inflammatory microglial phenotype via LPAR5 that couples to protein kinase D (PKD)-mediated pathways. Using the xCELLigence system and time-lapse microscopy, we investigated the migrational response of microglial cells. Different M1 and M2 markers were analyzed by confocal microscopy, flow cytometry, and immunoblotting. Using qPCR and ELISA, we studied the expression of migratory genes and quantitated the secretion of pro-inflammatory cytokines and chemokines, respectively. Different transcription factors that promote the regulation of pro-inflammatory genes were analyzed by western blot. Reactive oxygen species (ROS) and nitric oxide (NO) production, phagocytosis, and microglial cytotoxicity were determined using commercially available assay kits. LPA induces MAPK family and AKT activation and pro-inflammatory transcription factors' phosphorylation (NF-κB, c-Jun, STAT1, and STAT3) that were inhibited by both LPAR5 and PKD family antagonists. LPA increases migratory capacity, induces secretion of pro-inflammatory cytokines and chemokines and expression of M1 markers, enhances production of ROS and NO by microglia, and augments cytotoxicity of microglial cell-conditioned medium towards neurons. The PKD family inhibitor blunted all of these effects. We propose that interference with this signaling axis could aid in the development of new therapeutic approaches to control neuroinflammation under conditions of overshooting LPA production. In the present study, we show that inflammatory LPA levels increased the migratory response of microglia and promoted a pro-inflammatory phenotype via the LPAR5/PKD axis. Interference with this signaling axis reduced microglial migration, blunted microglial cytotoxicity, and abrogated the expression and secretion of pro-inflammatory mediators.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid