Merck
  • Differential regulation of synaptic AP-2/clathrin vesicle uncoating in synaptic plasticity.
Differential regulation of synaptic AP-2/clathrin vesicle uncoating in synaptic plasticity.
Scientific reports (2017-11-19)
Ermes Candiello, Ratnakar Mishra, Bernhard Schmidt, Olaf Jahn, Peter Schu
ABSTRACT
AP-1/σ1B-deficiency causes X-linked intellectual disability. AP-1/σ1B -/- mice have impaired synaptic vesicle recycling, fewer synaptic vesicles and enhanced endosome maturation mediated by AP-1/σ1A. Despite defects in synaptic vesicle recycling synapses contain two times more endocytic AP-2 clathrin-coated vesicles. We demonstrate increased formation of two classes of AP-2/clathrin coated vesicles. One which uncoats readily and a second with a stabilised clathrin coat. Coat stabilisation is mediated by three molecular mechanisms: reduced recruitment of Hsc70 and synaptojanin1 and enhanced μ2/AP-2 phosphorylation and activation. Stabilised AP-2 vesicles are enriched in the structural active zone proteins Git1 and stonin2 and synapses contain more Git1. Endocytosis of the synaptic vesicle exocytosis regulating Munc13 isoforms are differentially effected. Regulation of synaptic protein endocytosis by the differential stability of AP-2/clathrin coats is a novel molecular mechanism of synaptic plasticity.