Merck
  • Volatile Methylsiloxanes and Organophosphate Esters in the Eggs of European Starlings (Sturnus vulgaris) and Congeneric Gull Species from Locations across Canada.
Volatile Methylsiloxanes and Organophosphate Esters in the Eggs of European Starlings (Sturnus vulgaris) and Congeneric Gull Species from Locations across Canada.
Environmental science & technology (2017-08-05)
Zhe Lu, Pamela A Martin, Neil M Burgess, Louise Champoux, John E Elliott, Enzo Baressi, Amila O De Silva, Shane R de Solla, Robert J Letcher
ABSTRACT
Volatile methylsiloxanes (VMSs) and organophosphate esters (OPEs) are two suites of chemicals that are of environmental concern as organic contaminants, but little is known about the exposure of wildlife to these contaminants, particularly in birds, in terrestrial and aquatic ecosystems. The present study investigates the spatial distributions of nine cyclic and linear VMSs and 17 OPEs in the eggs of European starlings (Sturnus vulgaris) and three congeneric gull species (i.e., herring gull (Larus argentatus), glaucous-winged gull (L. glaucescens), and California gull (L. californicus)) from nesting sites across Canada. ∑VMS concentrations for all bird eggs were dominated by decamethylcyclopentasiloxane (D5), dodecamethylcyclohexasiloxane (D6), and octamethylcyclotetrasiloxane (D4). With European starlings, birds breeding adjacent to landfill sites had eggs containing significantly greater ∑VMS concentrations (median: 178 ng g
Sigma-Aldrich
Tripropyl phosphate, 99%
Sigma-Aldrich
Ethyltriphenylphosphonium bromide, 99%
Supelco
TCPP, mixture of isomers, PESTANAL®, analytical standard
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Dodecamethylpentasiloxane, 97%