Merck
  • Nucleocytoplasmic shuttling of bovine papillomavirus E1 helicase downregulates viral DNA replication in S phase.
Nucleocytoplasmic shuttling of bovine papillomavirus E1 helicase downregulates viral DNA replication in S phase.
Journal of virology (2006-10-13)
Chiung-Yueh Hsu, Francisca Mechali, Catherine Bonne-Andrea
ABSTRACT
The papillomavirus E1 protein is essential for the initiation of viral replication. We previously showed that the bovine papillomavirus E1 protein is unstable and becomes resistant to ubiquitin-mediated degradation when tightly bound to cyclin E-cyclin-dependent kinase 2 (Cdk2) before the start of DNA synthesis. However, neither the protection nor the targeted degradation of E1 appears to depend on its phosphorylation by Cdk. Here, we report that Cdk phosphorylation of E1 is also not a prerequisite for the initiation of viral DNA replication either in vitro or in vivo. Nevertheless, we found that phosphorylation of one Cdk site, Ser283, abrogates E1 replicative activity only in a cellular context. We show that this site-specific phosphorylation of E1 drives its export from the nucleus and promotes its continuous nucleocytoplasmic shuttling. In addition, we find that E1 shuttling occurs in S phase, when cyclin A-Cdk2 is activated. E1 interacts with the active cyclin A-Cdk2 complex and is phosphorylated on Ser283 by this kinase. These data suggest that the phosphorylation of E1 on Ser283 is a negative regulatory event that is involved in preventing the amplification of viral DNA during S phase. This finding reveals a novel facet of E1 regulation that could account for the variations of the viral replication capacity during different cell cycle phases, as well as in different stages of the viral cycle.