Merck
  • Pollutant-induced cell death and reactive oxygen species accumulation in the aerial roots of Chinese banyan (Ficus microcarpa).
Pollutant-induced cell death and reactive oxygen species accumulation in the aerial roots of Chinese banyan (Ficus microcarpa).
Scientific reports (2016-11-03)
Nan Liu, Ce Cao, Zhongyu Sun, Zhifang Lin, Rufang Deng
ABSTRACT
Industrial pollutants induce the production of toxic reactive oxygen species (ROS) such as O2.-, H2O2, and ·OH in plants, but they have not been well quantified or localized in tissues and cells. This study evaluated the pollutant- (HSO3-, NH4NO3, Al3+, Zn2+, and Fe2+) induced toxic effects of ROS on the aerial roots of Chinese banyan (Ficus microcarpa). Root cell viability was greatly reduced by treatment with 20 mM NaHSO3, 20 mM NH4NO3, 0.2 mM AlCl3, 0.2 mM ZnSO4, or 0.2 mM FeSO4. Biochemical assay and histochemical localization showed that O2.- accumulated in roots in response to pollutants, except that the staining of O2.- under NaHSO3 treatment was not detective. Cytochemical localization further indicated that the generated O2.- was present mainly in the root cortex, and pith cells, especially in NH4NO3- and FeSO4-treated roots. The pollutants also caused greatly accumulated H2O2 and ·OH in aerial roots, which finally resulted in lipid peroxidation as indicated by increased malondialdehyde contents. We conclude that the F. microcarpa aerial roots are sensitive to pollutant-induced ROS and that the histochemical localization of O2.- via nitrotetrazolium blue chloride staining is not effective for detecting the effects of HSO3- treatment because of the treatment's bleaching effect.
Sigma-Aldrich
XTT sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Benzenesulfonic acid monohydrate, 97%