Merck
  • MicroRNA-122 dependent binding of Ago2 protein to hepatitis C virus RNA is associated with enhanced RNA stability and translation stimulation.
MicroRNA-122 dependent binding of Ago2 protein to hepatitis C virus RNA is associated with enhanced RNA stability and translation stimulation.
PloS one (2013-02-14)
K Dominik Conrad, Florian Giering, Corinna Erfurth, Angelina Neumann, Carmen Fehr, Gunter Meister, Michael Niepmann
ABSTRACT
Translation of Hepatitis C Virus (HCV) RNA is directed by an internal ribosome entry site (IRES) in the 5'-untranslated region (5'-UTR). HCV translation is stimulated by the liver-specific microRNA-122 (miR-122) that binds to two binding sites between the stem-loops I and II near the 5'-end of the 5'-UTR. Here, we show that Argonaute (Ago) 2 protein binds to the HCV 5'-UTR in a miR-122-dependent manner, whereas the HCV 3'-UTR does not bind Ago2. miR-122 also recruits Ago1 to the HCV 5'-UTR. Only miRNA duplex precursors of the correct length stimulate HCV translation, indicating that the duplex miR-122 precursors are unwound by a complex that measures their length. Insertions in the 5'-UTR between the miR-122 binding sites and the IRES only slightly decrease translation stimulation by miR-122. In contrast, partially masking the miR-122 binding sites in a stem-loop structure impairs Ago2 binding and translation stimulation by miR-122. In an RNA decay assay, also miR-122-mediated RNA stability contributes to HCV translation stimulation. These results suggest that Ago2 protein is directly involved in loading miR-122 to the HCV RNA and mediating RNA stability and translation stimulation.