- Transcriptional response of marine medaka (Oryzias javanicus) on exposure to toxaphene.
Transcriptional response of marine medaka (Oryzias javanicus) on exposure to toxaphene.
Differential gene expression profiles were established from the head and liver tissues of the marine medaka fish (Oryzias javanicus) after its exposure to toxaphene, a persistent organic pollutant and insecticide, using differential display polymerase chain reaction. Twenty-seven differentially expressed genes were identified, which were associated with the cytoskeleton, development, metabolism, nucleic acid/protein binding, and signal transduction. Among these genes, those encoding molecular biomarkers known to be involved in metabolism, ATP hydrolysis, and protein regulation were strongly induced at the transcription level, and genes encoding one structural protein subunit or involved in lipid metabolism were strongly downregulated. The same trends in gene expression changes were observed with real-time PCR analysis of 12 selected clones. The genes identified could be used as molecular biomarkers of biological responses to polychlorinated camphene contamination in aquatic environments.