Merck
  • Successful treatment of recurrent epithelial ingrowth associated with interface fluid syndrome, flap necrosis, and epithelial defects following LASIK.
Successful treatment of recurrent epithelial ingrowth associated with interface fluid syndrome, flap necrosis, and epithelial defects following LASIK.
Journal of refractive surgery (Thorofare, N.J. : 1995) (2010-06-15)
Saba J Elderkin, Randy J Epstein, Dianna L Seldomridge
ABSTRACT
to report the successful treatment of two patients who developed flap necrosis preceded by recurrent epithelial ingrowth and interface fluid syndrome after LASIK. patient 1 was treated with epithelial debridement and flap suturing. Patient 2 was initially treated with epithelial debridement and flap suturing, but developed recurrent epithelial ingrowth in the right eye and 2 weeks later in the left eye. patient 1 developed diffuse interface fluid accumulation in the left eye after epithelial debridement and flap suturing and was treated with timolol meleate 0.5% solution and methazolamide. The interface fluid resolved and the cornea and flap became clear. Slit-lamp examination identified a small area of epithelial ingrowth recurrence, which has remained stable for 3 years. Patient 2 was successfully retreated with epithelial debridement followed by fibrin tissue adhesive application. Five months after debridement and fibrin tissue adhesive, no recurrence of epithelial ingrowth or interface fluid accumulation was noted. epithelial ingrowth and interface fluid syndrome may be associated with secondary flap necrosis following LASIK, which can be effectively treated with debridement and flap suturing or fibrin tissue adhesive application.