Merck
  • Urinary levels of the tobacco-specific carcinogen N'-nitrosonornicotine and its glucuronide are strongly associated with esophageal cancer risk in smokers.
Urinary levels of the tobacco-specific carcinogen N'-nitrosonornicotine and its glucuronide are strongly associated with esophageal cancer risk in smokers.
Carcinogenesis (2011-07-08)
Jian-Min Yuan, Aleksandar D Knezevich, Renwei Wang, Yu-Tang Gao, Stephen S Hecht, Irina Stepanov
ABSTRACT
N'-Nitrosonornicotine (NNN) and 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) are tobacco-specific nitrosamines. NNN and NNK can induce cancers of the esophagus and lung, respectively, in laboratory animals, but data on human esophageal cancer are lacking. The association between levels of NNN and 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol (NNAL), an NNK metabolite, in urine samples collected before diagnosis and risk of esophageal cancer was examined in 77 patients with esophageal cancer and 223 individually matched controls, all current smokers, from a cohort of 18244 Chinese men in Shanghai, China, followed from 1986 to 2008. Urinary total NNN (free NNN plus NNN-N-glucuronide) was significantly higher, whereas the percentage of its detoxification product NNN-N-glucuronide was significantly lower in cases than controls. Odds ratios (95% confidence intervals) of esophageal cancer for the second and third tertiles of total NNN were 3.99 (1.25-12.7) and 17.0 (3.99-72.8), respectively, compared with the first tertile after adjustment for urinary total NNAL and total cotinine and smoking intensity and duration (P(trend) < 0.001). The corresponding figures for the percentage of NNN-N-glucuronides were 0.37 (0.17-0.80) and 0.27 (0.11-0.62) (P(trend) = 0.001). Urinary total NNN and the percentage of NNN-N-glucuronides almost completely accounted for the observed association for urinary total NNAL (free NNAL plus its glucuronides), urinary total cotinine and smoking intensity with esophageal cancer risk. These findings along with results of previous studies in laboratory animals support a significant and unique role of NNN in esophageal carcinogenesis in humans.
Product Description
Supelco
(±)-N′-Nitrosonornicotine (NNN) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Supelco
N′-Nitrosonornicotine, analytical standard
Pricing and availability is not currently available.