Merck
  • Blocking glutamate-mediated signalling inhibits human melanoma growth and migration.
Blocking glutamate-mediated signalling inhibits human melanoma growth and migration.
Experimental dermatology (2012-11-23)
Zhiqi Song, Chun-Di He, Jing Liu, Changkai Sun, Ping Lu, Lili Li, Lili Gao, Yi Zhang, Yanni Xu, Lujuan Shan, Yuejian Liu, Wei Zou, Yuan Zhang, Haiqin Gao, Wenting Gao
ABSTRACT
Glutamate is an excitatory neurotransmitter that has been shown to regulate the proliferation, migration and survival of neuronal progenitors in the central nervous system through its action on metabotropic and ionotropic glutamate receptors (GluRs). Antagonists of ionotropic GluRs have been shown to cause a rapid and reversible change in melanocyte dendritic morphology, which is associated with the disorganization of actin and tubulin microfilaments in the cytoskeleton. Intracellular expression of microtubule-associated protein (MAP) 2a affects the assembly, stabilization and bundling of microtubules in melanoma cells; stimulates the development of dendrites; and suppresses melanoma cell migration and invasion. In this study, we investigated the relationship between glutamate-mediated signalling and microtubules, cell dendritic morphology and melanoma cell motility. We found that metabotropic GluR1 and N-methyl-d-aspartate receptor antagonists increased dendritic branching and inhibited the motility, migration and proliferation of melanoma cells. We also demonstrated that the invasion and motility of melanoma cells are significantly inhibited by the combination of increased expression of MAP2a and either metabotropic GluR1 or N-methyl-d-aspartate receptor antagonists. Moreover, the blockade of glutamate receptors inhibited melanoma growth in vivo. Collectively, these results demonstrate the importance of glutamate signalling in human melanoma and suggest that the blockade of glutamate receptors is a promising novel therapy for treating melanoma.
Sigma-Aldrich
Anti-NMDAR2A Antibody, Chemicon®, from mouse