Merck
  • FER regulates endosomal recycling and is a predictor for adjuvant taxane benefit in breast cancer.
FER regulates endosomal recycling and is a predictor for adjuvant taxane benefit in breast cancer.
Cell reports (2022-04-07)
Sandra Tavares, Nalan Liv, Milena Pasolli, Mark Opdam, Max A K Rätze, Manuel Saornil, Lilian M Sluimer, Rutger C C Hengeveld, Robert van Es, Erik van Werkhoven, Harmjan Vos, Holger Rehmann, Boudewijn M T Burgering, Hendrika M Oosterkamp, Susanne M A Lens, Judith Klumperman, Sabine C Linn, Patrick W B Derksen
ABSTRACT
Elevated expression of non-receptor tyrosine kinase FER is an independent prognosticator that correlates with poor survival of high-grade and basal/triple-negative breast cancer (TNBC) patients. Here, we show that high FER levels are also associated with improved outcomes after adjuvant taxane-based combination chemotherapy in high-risk, HER2-negative patients. In TNBC cells, we observe a causal relation between high FER levels and sensitivity to taxanes. Proteomics and mechanistic studies demonstrate that FER regulates endosomal recycling, a microtubule-dependent process that underpins breast cancer cell invasion. Using chemical genetics, we identify DCTN2 as a FER substrate. Our work indicates that the DCTN2 tyrosine 6 is essential for the development of tubular recycling domains in early endosomes and subsequent propagation of TNBC cell invasion in 3D. In conclusion, we show that high FER expression promotes endosomal recycling and represents a candidate predictive marker for the benefit of adjuvant taxane-containing chemotherapy in high-risk patients, including TNBC patients.
Sigma-Aldrich
Anti-DCTN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Tubulin, GST tagged human, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
DAPI, for nucleic acid staining