Merck
  • Synthetic DNA spike-ins (SDSIs) enable sample tracking and detection of inter-sample contamination in SARS-CoV-2 sequencing workflows.
Synthetic DNA spike-ins (SDSIs) enable sample tracking and detection of inter-sample contamination in SARS-CoV-2 sequencing workflows.
Nature microbiology (2021-12-16)
Kim A Lagerborg, Erica Normandin, Matthew R Bauer, Gordon Adams, Katherine Figueroa, Christine Loreth, Adrianne Gladden-Young, Bennett M Shaw, Leah R Pearlman, Daniel Berenzy, Hannah B Dewey, Susan Kales, Sabrina T Dobbins, Erica S Shenoy, David Hooper, Virginia M Pierce, Kimon C Zachary, Daniel J Park, Bronwyn L MacInnis, Ryan Tewhey, Jacob E Lemieux, Pardis C Sabeti, Steven K Reilly, Katherine J Siddle
ABSTRACT
The global spread and continued evolution of SARS-CoV-2 has driven an unprecedented surge in viral genomic surveillance. Amplicon-based sequencing methods provide a sensitive, low-cost and rapid approach but suffer a high potential for contamination, which can undermine laboratory processes and results. This challenge will increase with the expanding global production of sequences across a variety of laboratories for epidemiological and clinical interpretation, as well as for genomic surveillance of emerging diseases in future outbreaks. We present SDSI + AmpSeq, an approach that uses 96 synthetic DNA spike-ins (SDSIs) to track samples and detect inter-sample contamination throughout the sequencing workflow. We apply SDSIs to the ARTIC Consortium's amplicon design, demonstrate their utility and efficiency in a real-time investigation of a suspected hospital cluster of SARS-CoV-2 cases and validate them across 6,676 diagnostic samples at multiple laboratories. We establish that SDSI + AmpSeq provides increased confidence in genomic data by detecting and correcting for relatively common, yet previously unobserved modes of error, including spillover and sample swaps, without impacting genome recovery.
Sigma-Aldrich
KOD Hot Start Master Mix, Ready-to-use 2X mixture, containing KOD Hot Start DNA Polymerase, two monoclonal antibodies, ultrapure deoxynucleotides, and reaction buffer with MgSO4, optimized for convenient high fidelity PCR., ready-to-use solution, suitable for PCR