- 8-Amino-6-Methoxyquinoline-Tetrazole Hybrids: Impact of Linkers on Antiplasmodial Activity.
8-Amino-6-Methoxyquinoline-Tetrazole Hybrids: Impact of Linkers on Antiplasmodial Activity.
Molecules (Basel, Switzerland) (2021-09-29)
Patrick Hochegger, Johanna Dolensky, Werner Seebacher, Robert Saf, Marcel Kaiser, Pascal Mäser, Robert Weis
PMID34577001
ABSTRACT
A new series of compounds was prepared from 6-methoxyquinolin-8-amine or its N-(2-aminoethyl) analogue via Ugi-azide reaction. Their linkers between the quinoline and the tert-butyltetrazole moieties differ in chain length, basicity and substitution. Compounds were tested for their antiplasmodial activity against Plasmodium falciparum NF54 as well as their cytotoxicity against L-6-cells. The activity and the cytotoxicity were strongly influenced by the linker and its substitution. The most active compounds showed good activity and promising selectivity.