Merck
Extended-spectrum cephalosporinase in Acinetobacter baumannii.
Antimicrobial agents and chemotherapy (2010-06-16)
José-Manuel Rodríguez-Martínez, Patrice Nordmann, Esthel Ronco, Laurent Poirel
ABSTRACT
An AmpC-type beta-lactamase conferring high-level resistance to expanded-spectrum cephalosporins and monobactams was characterized from an Acinetobacter baumannii clinical isolate. This class C beta-lactamase (named ADC-33) possessed a Pro210Arg substitution together with a duplication of an Ala residue at position 215 (inside the Omega-loop) compared to a reference AmpC cephalosporinase from A. baumannii. ADC-33 hydrolyzed ceftazidime, cefepime, and aztreonam at high levels, which allows the classification of this enzyme as an extended-spectrum AmpC (ESAC). Site-directed mutagenesis confirmed the role of both substitutions in its ESAC property.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis